Fraction collectors, FC 204
Lieferant: INTERCHIM
FSQ610
INTEFSQ610EA 0 EUR
INTEFSQ610
Fraction collectors, FC 204
Fraktionssammler
The FC 204 is the ideal large bed fraction collector, capable of handling a wide variety of collection vessels including microplates, vials and tubes. The built in key-pad makes setup and operation simple, capable of interfacing with any HPLC system via contact signals. In peak mode, the collector monitors a detector signal and identifies peaks using either an adaptive-slope algorithm or a specified millivolt threshold.
  • Stand-alone, with an easy-to-use keypad or can be controlled TRILUTION® Software via PC
  • Multicolumn adapters for collecting up to 18 separate channels
  • Operating modesm: Drop, manual, peak + drop, peak + time, time
  • 3-way diverter valve prevents contamination of collected fractions