Pump tubing, Viton®
Lieferant: ISMATEC PART OF COLE PARMER
06519-08
ISMC06519-08EA 0 EUR
ISMC06519-08
Pump tubing, Viton®
Schläuche Transferschlauch
Fluoroelastomer, black
  • High chemical resistance
  • Low gas permeability
  • Wide temperature range
The special tubing for concentrated acids and corrosive solvents.
Temperature range: -51 to +204 °C
Hardness (Shore durometer A): 60
Density: 1,90