BenchBin™ Benchtop Biohazard Bin
Lieferant: ISMATEC PART OF COLE PARMER
14003-04
ISMC14003-04EA 0 EUR
ISMC14003-04
BenchBin™ Benchtop Biohazard Bin
Behälter Abfallbehälter
The BenchBin™ is a compact, convenient, and safe biohazard bin for the benchtop or workstation.
  • Push to open/close lid keeps waste materials covered
  • Use with 216×279 mm disposal bags
  • Compatible with autoclavable biohazard bags
Benchtop biohazard bin only measures 180L×130W×180H mm taking up minimal countertop space. Removable cover makes it quick and easy to empty contents.