Heating tapes, TeMS2
Lieferant: ISOPAD
411376-000 755718-000 533964-000 184778-000 584216-000 067444-000 458786-000 124922-000 927204-000
ISOP411376-000EA 276.5 EUR
ISOP411376-000 454-2191 454-2192 ISOP184778-000 ISOP584216-000 ISOP067444-000 ISOP458786-000 ISOP124922-000 ISOP927204-000
Heating tapes, TeMS2
Heizgeräte
Isopad IT-TeMS is a preterminated and PTFE insulated flexible heating tape.
  • Temperature resistant up to 260 °C
  • Non hazardous and used in ordinary areas
  • Parallel arrangement ensures a wide heat transfer surface
IT-TeMS2 is used in applications ranging from simple frost protection to temperature maintenance on short pipework and apparatus as well as for general container heating applications. TeMS4 or TeMS6 tapes can be used when more power is required.
Neuer Artikel bei VWR!
Bestellen
TECHNISCHE DATEN
Environmentally Preferable