PeelASeal Foil Super Heat Sealing Film 1 Trial Roll 5m x 115mm Peelable from all plate types Seal integrity range: -80 °C to 110 °C Good solvent resistance including DMSO Autoclavable (121°C) 1 * 1 Rolle
Lieferant: iST Scientific Ltd
PeelASeal Foil Super Heat Sealing Film 1 Trial Roll 5m x 115mm Peelable from all plate types Seal integrity range: -80 °C to 110 °C Good solvent resistance including DMSO Autoclavable (121°C) 1 * 1 Rolle