Adeninsulfat Dihydat ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Aminopurine sulphate dihydrate, Adenine hemisulphate dihydrate, 7H-Purin-6-amine hemisulphate dihydrate, 9H-Purin-6-amine hemisulphate dihydrate

J10450.22
J10450.22EA 163 EUR
J10450.22
Adeninsulfat Dihydat ≥97%
Adeninsulfat Dihydat
Formel: (C₅H₅N₅)₂·H₂SO₄·2H₂O
Molecular Weight: 404,37 g/mol
Schmelzpunkt: ∼ 210 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00213655
CAS-Nummer: 321-30-2
EINECS: 206-286-5
Merck Index: 14,152

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR