Agar, ultrarein für die Bakteriologie

Lieferant: Thermo Fisher Scientific

Synonyme: Agar Agar

J10906.P5
J10906.P5EA 393 EUR
J10906.P5
Agar, ultrarein für die Bakteriologie
Agar

Agar is used extensively as a gelling agent in the preparation of bacteriological culture media.


Its main attributes are superior transparency, very reliable reproducibility and absence of inhibitors capable of hindering optimal development of microorganisms. Extraneous matter reduced to a minimum.

Schmelzpunkt: 90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081288
CAS-Nummer: 9002-18-0
EINECS: 232-658-1
Merck Index: 14,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Gel Point (1.5%) 32 - 38 °C
Assay Gel Strength (1.5%) 550 - 950 gm/cm²
Assay Gel pH (1.5%) after Autoclaving 6.0 - 7.5
Ash ≤6.5%
Residual Water ≤12%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR