Citronensäure, wasserfrei 99,5-100,05% (Trockenmasse)
Lieferant: Thermo Fisher Scientific
J13729.A1
J13729.36
J13729.A1EA
87.9
EUR
J13729.A1
J13729.36
Citronensäure, wasserfrei 99,5-100,05% (Trockenmasse)
Spezifikation Testergebnisse
Assay | 99.5 - 100.5% (dry basis) |
Assay, Readily Carbonisable Substances | Passes Test |
Identification (FTIR) | Conforms |
Residue on Ignition | ≤0.1% |
Impurity Content, Oxalate | Passes Test |
Impurity Content, Sulphate (SO₄) | Passes test |
Water Content (Karl Fischer Titration) | ≤1.0% |
Heavy Metals (Pb) | ≤10 µg/gm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...