2'-Deoxyguanosine monohydrate 98+% (Trockenmasse)
Lieferant: Thermo Scientific
Synonyme:
2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one hydrate
J14304.09
J14304.09EA
646
EUR
J14304.09
2'-Deoxyguanosine monohydrate 98+% (Trockenmasse)
2'-Deoxyguanosine monohydrate
Formel:
C₁₀H₁₃N₅O₄·H₂O Molecular Weight: 285,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150760 CAS-Nummer: 312693-72-4 EINECS: 213-505-8 |
Spezifikation Testergebnisse
Form | White crystalline powder |
Assay (unspecified) | ≥ 98% (dry basis) |
Assay (special): Absorbance Ratios: A250/A260, A280/A260 | 1.12 to 1.18, 0.65 to 0.69 (pH 7.0) |
Assay (special): Extinction Coefficient | 13.7 X 103 ± 4% (pH 7.0) |
Identification (FTIR) | Nujol mull matches reference scan |
Water Content (Karl Fischer Titration) | ≤ 7.0% |
lambda-max (UV-Vis) | 253 ± 2 nm (pH 7.0) |
Thin Layer Chromatography | Homogenous |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...