EDTA Tetranatriumsalz Dihydrat 99-101%, ultrarein

Lieferant: Thermo Fisher Scientific
Gefahr

J15700.A1 J15700.C4
J15700.A1EA 73.1 EUR
J15700.A1 J15700.C4
EDTA Tetranatriumsalz Dihydrat 99-101%, ultrarein
EDTA Tetranatriumsalz Dihydrat
Formel: C₁₀H₁₂N₂Na₄O₈·2H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150026
CAS-Nummer: 10378-23-1

Bestellen

Spezifikation Testergebnisse

Form White crystalline powder
Assay (unspecified) 99 to 101% (as is)
Identification (FTIR) Conforms
pH (1%, H2O) 10.7 to 11.7
pH (5%, H2O) 10.7 to 12.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR