Jenner's stain, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Eosin−Methylene Blue

J1625-25G
J1625-25GEA 202 EUR
J1625-25G
Jenner's stain, Sigma-Aldrich®
Jenner's stain

Jenner's stain, suitable for blood stain (Eosin-Methylene Blue)

Lagertemperatur: Raumtemperatur
MDL: MFCD00081733
CAS-Nummer: 62851-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR