HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure) ≥99% (Trockenmasse), Pulver, ultrarein
Lieferant: Thermo Fisher Scientific
Synonyme:
HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
J16926.A1
J16926.22
J16926.A7
J16926.A1EA
437
EUR
J16926.A1
J16926.22
J16926.A7
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure) ≥99% (Trockenmasse), Pulver, ultrarein
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
Formel:
C₈H₁₈N₂O₄S Molecular Weight: 238.31 g/mol Schmelzpunkt: ∼ 236 °C (Zers.) Dichte: 1.448 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006158 CAS-Nummer: 7365-45-9 EINECS: 230-907-9 Merck Index: 14,04654 |
|
Spezifikation Testergebnisse
Assay (Unspecified) | ≥99.0% (Dry Basis) |
Identification (FTIR) | Conforms |
pKa (20 °C) | 7.55 ±0.15 |
A (250 nm, 1 M, 1 cm) | ≤0.2 |
Loss on Drying | ≤0.5% |
Residue on Ignition | ≤0.1% |
Chloride (Cl) | ≤0.005% |
Phosphate (PO₄) | ≤0.001% |
Sulphate (SO₄) | ≤0.01% |
Iron (Fe) | ≤5 ppm |
Heavy Metals (Pb) | ≤10 ppm |
Assay (Unspecified) Absorbance (250 nm, 1 M, 1 cm) | ≤0.2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...