L(+)-Histidin Hydrochlorid Monohydrat 98+%

Lieferant: Thermo Scientific

Synonyme: His HCl Monohydrat, (S)-(+)-Histidin Hydrochlorid Monohydrat, (2S)-(+)-Histidin Hydrochlorid Monohydrat, (S)-(+)-2-Amino-3-(4-imidazoyl)propansäure Hydrochlorid Monohydrat, L-3-Imidazol-4-ylalanin Hydrochlorid Monohydrat, H-His-OH.HCl.H2O

J17080.A1
J17080.A1EA 343 EUR
J17080.A1
L(+)-Histidin Hydrochlorid Monohydrat 98+%
L(+)-Histidin Hydrochlorid Monohydrat
Formel: C₆H₉N₃O₂·HCl·H₂O
Molecular Weight: 209,63 g/mol
Schmelzpunkt: 240 °C (Zersetzung)
Dichte: 1,485 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151027
CAS-Nummer: 5934-29-2
EINECS: 211-438-9
Merck Index: 14,04720
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (unspecified) - Ammonium (NH₄) ≤0.02%
Assay (unspecified) ≥98.0%
Identification (FTIR) Conforms
Residue on Ignition ≤0.10%
Impurity Content Sulphate (SO₄) ≤0.03%
Loss on Drying ≤0.3%
Optical Rotation (c= in 6 N HCl) +9.2 to 10.6°
Thin Layer Chromatography Homogeneous
Arsenic (As₂O₃) ≤1 ppm
Iron (Fe) ≤30 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR