Tris-HCl, 1M solution, pH 8,0, Molecular Biology Grade, Ultrapure
Lieferant: Thermo Fisher Scientific
J22638.AE
J22638.AP
J22638.AEEA
37.7
EUR
J22638.AE
J22638.AP
Tris-HCl, 1M solution, pH 8,0, Molecular Biology Grade, Ultrapure
Puffer
Klonpuffer
1M solution of Tris, Ultrapure in high purity distilled H₂O, adjusted to pH 8,0.