Tris(hydroxymethyl)aminomethan (TRIS, Trometamol) ≥98,0% (Trockenmasse)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Aminomethylidintrimethanol, Tris(hydroxymethyl)methylamin, THAM, 2-Amino-2-(hydroxymethyl)propan-1,3-diol, Tromethamin, TRIS Base, 2-Amino-2-(hydroxymethyl)-1,3-propandiol

J22675.A1 J22675.36
J22675.A1EA 95.1 EUR
J22675.A1 J22675.36
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol) ≥98,0% (Trockenmasse)
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Formel: NH₂C(CH₂OH)₃
Molecular Weight: 121.14 g/mol
Siedepunkt: 219…220 °C (10 mmHg)
Schmelzpunkt: 171…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004679
CAS-Nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 14,09772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (Dry Basis) ≥98.0%
Identification (FTIR) Conforms
Loss on Drying ≤1.5%
pH (5%, H₂O) 10.0 - 11.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR