4-Nitrophenyl-ɑ-L-fucopyranoside
Lieferant: Thermo Scientific
J34131.03
J34131.03EA
528
EUR
J34131.03
4-Nitrophenyl-ɑ-L-fucopyranoside
4-Nitrophenyl-alpha-L-fucopyranoside
Formel:
C₁₂H₁₅NO₇ Molecular Weight: 285,25 g/mol Schmelzpunkt: 194…198 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00063697 CAS-Nummer: 10231-84-2 EINECS: 233-553-3 |
Spezifikation Testergebnisse
Identification (FTIR) | Conforms |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...