N,N-Dimethylacetamid ≥99,5% (durch GC), BAKER ANALYZED®, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
J372-07EA
329.09
EUR
J372-07
N,N-Dimethylacetamid ≥99,5% (durch GC), BAKER ANALYZED®, J.T.Baker®
N,N-Dimethylacetamid
Formel:
CH₃CON(CH₃)₂ Siedepunkt: 166,1 °C (1013 hPa) Schmelzpunkt: –20 °C Dichte: 0,937 g/cm³ (20 °C) Flammpunkt: 70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008686 CAS-Nummer: 127-19-5 EINECS: 204-826-4 UN: 2810 ADR: 6.1,III Merck Index: 13,03254 |
|
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use |
|
Assay (CH₃CON(CH₃)₂) (by GC) | ≥99.5 % |
Density (g/mL) at 25°C | 0.931 - 0.936 |
Residue after Ignition | ≤0.005 % |
pH of 20% Solution at 25°C | 4.0 - 7.0 |
Water (H₂O) | ≤0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...