4-(Dimethylamino)benzaldehyd, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
J418-05EA
232.02
EUR
J418-05
4-(Dimethylamino)benzaldehyd, BAKER ANALYZED® ACS, J.T.Baker®
4-(Dimethylamino)benzaldehyd
Lot analysis on label.
Formel:
(CH₃)₂NC₆H₄CHO Siedepunkt: 176…177 °C (1013 hPa) Schmelzpunkt: 72 °C Dichte: 1,0254 g/cm³ (99.9 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003381 CAS-Nummer: 100-10-7 EINECS: 202-819-0 Merck Index: 13,03257 |
|
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | 73 - 75 °C |
For Laboratory,Research,or Manufacturing Use | 73 - 75 °C |
Melting Point | 73 - 75 °C |
Solubility in HCl | Passes Test |
Solubility in Alcohol | Passes Test |
Color of HCl Solution | Passes Test |
Color of Alcohol Solution(APHA) | ≤60 |
Residue after Ignition | ≤0.10 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...