Thiamine nitrate
Lieferant: Thermo Fisher Scientific
Synonyme:
Vitamin B1 Nitrate
J60014.09
J60014.14
J60014.09EA
41.5
EUR
J60014.09
J60014.14
Thiamine nitrate
Thiamine nitrate
CAS No.: 532-43-4
Molecular Formula: C12H17N5O4S
Formula Weight: 327.36
Physical Form: Powder
Appearance: White
Melting Point: 190°
MDL No.: MFCD00036330
Formel:
C₁₂H₁₇N₅O₄S Schmelzpunkt: 190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036330 CAS-Nummer: 532-43-4 EINECS: 208-537-4 Merck Index: 14,9295 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...