Thiamine nitrate

Lieferant: Thermo Fisher Scientific

Synonyme: Vitamin B1 Nitrate

J60014.09 J60014.14
J60014.09EA 41.5 EUR
J60014.09 J60014.14
Thiamine nitrate
Thiamine nitrate

CAS No.: 532-43-4 Molecular Formula: C12H17N5O4S Formula Weight: 327.36 Physical Form: Powder Appearance: White Melting Point: 190° MDL No.: MFCD00036330

Formel: C₁₂H₁₇N₅O₄S
Schmelzpunkt: 190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036330
CAS-Nummer: 532-43-4
EINECS: 208-537-4
Merck Index: 14,9295

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR