Laemmli SDS sample buffer, reducing (4X)

Lieferant: Thermo Fisher Scientific

J60015.ACEA 76.5 EUR
J60015.AC J60015.AD
Laemmli SDS sample buffer, reducing (4X)
Puffer Elektrophorese-Puffer Beladungspuffer

Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Bestellen

TECHNISCHE DATEN

Environmentally Preferable
Aussehen Blue
Physical form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR