Transformation reagent, TB solution-II
Lieferant: Thermo Scientific
J60028.AK
J60028.AKEA
135
EUR
J60028.AK
Transformation reagent, TB solution-II
Nukleinsäure-Reagenzien
Transformations-Reagenzien
For transformation of competent cells.
Contains 250 mM potassium chloride, 15 mM calcium chloride, 10 mM HEPES, 55 mM manganese chloride, pH 6,5.
Applications: For transformation of competent cells.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...