N-Lauroylsarkosin Natriumsalz 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 95%
CAS No.: 137-16-6
Molecular Formula: C15H29NNaO3
Formula Weight: 293.39
Physical Form: Crystalline powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 44-47°
Beilstein Reference: 5322974
Merck Reference: 14,4368
MDL No.: MFCD00042728
Used in the cell lysis step of RNA purification. Anionic detergent
Formel:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa Schmelzpunkt: 44…47 °C Flammpunkt: >150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042728 CAS-Nummer: 137-16-6 EINECS: 205-281-5 UN: 2811 ADR: 6.1,II Merck Index: 14,4368 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...