Acriflavin Hydrochlorid

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,6-Diamino-10-methylacridinium chloride hydrochloride, Euflavine

J60048.14
J60048.14EA 51.8 EUR
J60048.14
Acriflavin Hydrochlorid
Acriflavin Hydrochlorid
Formel: C₂₇H₂₈Cl₄N₆
Molecular Weight: 578,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069039
CAS-Nummer: 8063-24-9
UN: 3077
ADR: 9,III
Merck Index: 14,123

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR