BES, 0.5 M buffer solutions

Lieferant: Thermo Fisher Scientific

J60058.AK
J60058.AKEA 76 EUR
J60058.AK
BES, 0.5 M buffer solutions
Puffer Biologische Puffer

CAS No.: 66992-27-6
Storage temperature: 2 to 8 °C
Physical form: Liquid
Appearance: Colourless
MDL No.: MFCD00067425

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR