TRIS Pufferlösung pH 8 (1 mol/l)

Lieferant: Thermo Fisher Scientific

J60080.AK
J60080.AKEA 82.3 EUR
J60080.AK
TRIS Pufferlösung pH 8 (1 mol/l)
Puffer Klonpuffer

CAS No.: 1185-53-1
Physical form: liquid
Appearance: colourless
MDL No.: MFCD00012590

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR