Fig tree Ficin (from Latex)
Lieferant: Thermo Fisher Scientific
J60136.14
J60136.14EA
242
EUR
J60136.14
Fig tree Ficin (from Latex)
Enzyme
Applications: Proteolytic enzyme from fig latex, that is a cysteine endopeptidase