Uridine-5'-diphosphate trisodium salt ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
UDP sodium salt
J60141.MC
J60141.MCEA
102
EUR
J60141.MC
Uridine-5'-diphosphate trisodium salt ≥98%
Uridine-5'-diphosphate trisodium salt
Purity: 98+%
CAS No.: 21931-53-3
Molecular Formula: C9H14N2Na3O12P2
Formula Weight: 470.11
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water to 100mM
MDL No.: MFCD00006524
Formel:
C₉H₁₄N₂Na₃O₁₂P₂ Molecular Weight: 470.11 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006524 CAS-Nummer: 21931-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...