Actinomycin D, Pulver
Lieferant: Thermo Scientific
J60148.X0
J60148.X0EA
120
EUR
J60148.X0
Actinomycin D, Pulver
Actinomycin D
Inhibits nulceic acid synthesis and potently induces apoptosis. Used in banding techniques to differentiate between different regions of chromosomes. Also used as a selection agent
Formel:
C₆₂H₈₆N₁₂O₁₆ Molecular Weight: 1255,5 g/mol Schmelzpunkt: 251…253 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005033 CAS-Nummer: 50-76-0 UN: 3462 ADR: 6.1,II Merck Index: 14,2800 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...