L-Kynurenine

Lieferant: Thermo Scientific

Synonyme: β-Anthraniloyl-L-alanine, (S)-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, Kynurenine

J60199.MF
J60199.MFEA 72.3 EUR
J60199.MF
L-Kynurenine
L-Kynurenine

CAS No.: 2922-83-0 Molecular Formula: C10H12N2O3 Formula Weight: 208.21 Physical Form: Crystalline Appearance: Pale yellow MDL No.: MFCD00069912


Applications: Intermediate in the breakdown of tryptophan

Formel: C₁₀H₁₂N₂O₃
Molecular Weight: 208,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069912
CAS-Nummer: 2922-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR