L(+)-Ornithin Hydrochlorid Reagenz für die Zellkultur
Lieferant: Thermo Fisher Scientific
CAS No.: 3184-13-2
Molecular Formula: C5H12N2O2.HCl
Formula Weight: 168.62
Physical Form: Crystalline powder
Appearance: White
Melting Point: ca 238° dec.
Beilstein Reference: 3625847
Merck Reference: 14,6874
Optical Rotation: +23° (c=5 in 5N HCl)
MDL No.: MFCD00064562
Formel:
C₅H₁₂N₂O₂·HCl Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064562 CAS-Nummer: 3184-13-2 EINECS: 221-678-6 Merck Index: 14,6874 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...