Kinase buffer II (5X)
Lieferant: Thermo Fisher Scientific
J60269.AE
J60269.AK
J60269.AEEA
46.8
EUR
J60269.AE
J60269.AK
Kinase buffer II (5X)
Puffer
Assay-Puffer
Physical form: Liquid
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...