N-Methyldopamine hydrochloride 95%
Lieferant: Thermo Fisher Scientific
J60306.03
J60306.03EA
235
EUR
J60306.03
N-Methyldopamine hydrochloride 95%
N-Methyldopamine hydrochloride
Purity: 95%
CAS No.: 62-32-8
Molecular Formula: C9H13NO2.HCl
Formula Weight: 203.67
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00035074
Applications: Dopamine agonist
Formel:
C₉H₁₃NO₂·HCl Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035074 CAS-Nummer: 62-32-8 EINECS: 200-528-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...