Tyrphostin A23 99%
Lieferant: Thermo Scientific
Synonyme:
2-(3,4-Dihydroxybenzylidene)malononitrile
J60308.MA
J60308.MAEA
120
EUR
J60308.MA
Tyrphostin A23 99%
Tyrphostin A23
Purity: 99%
CAS No.: 118409-57-7
Molecular Formula: C10H6N2O2
Formula Weight: 186.17
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Yellow to pale brown
Solubility: Soluble in DMSO, ethanol, or DMF
MDL No.: MFCD00133899
Applications: Potent inhibitor of EGF receptor kinase activity
Formel:
C₁₀H₆N₂O₂ Molecular Weight: 186,1 g/mol |
MDL:
MFCD00133899 CAS-Nummer: 118409-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...