Phosphate, 0,2 M buffer solution, pH 6,8
Lieferant: Thermo Fisher Scientific
J60325.K2
J60325.K2EA
101
EUR
J60325.K2
Phosphate, 0,2 M buffer solution, pH 6,8
Puffer
Biologische Puffer
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...