3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydrochloride TMB, TMB-HCl

J60332.03 J60332.06 J60332.88
J60332.03EA 73.5 EUR
J60332.03 J60332.06 J60332.88
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat 99+%
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat

Purity: 99+% CAS No.: 207738-08-7 Molecular Formula: C16H20N2.2HCl.xH2O Formula Weight: 313.27(anhy) Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Moisture and Light Sensitive Appearance: White to off-white MDL No.: MFCD00150104


Excellent Colourimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

Formel: C₁₆H₂₀N₂·2HCl·xH₂O
Lagertemperatur: Gefrierschrank
MDL: MFCD00150104
CAS-Nummer: 207738-08-7
EINECS: 264-769-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR