Astemizole ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(4-Fluorobenzyl)-2-(1-[4-methoxyphenethyl]piperidin-4-yl)aminobenzimidazole, astemizole
J60339.MF
J60339.MFEA
157
EUR
J60339.MF
Astemizole ≥99%
Astemizole
Purity: 99+%
CAS No.: 68844-77-9
Molecular Formula: C28H31FN4O
Formula Weight: 458.57
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 149-150°
MDL No.: MFCD00153919
Applications: Selective histamine H1 receptor antagonist
Formel:
C₂₈H₃₁FN₄O Molecular Weight: 458,57 g/mol Schmelzpunkt: 149…150 °C |
MDL:
MFCD00153919 CAS-Nummer: 68844-77-9 EINECS: 272-441-9 Merck Index: 14,856 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...