Chloropyramine hydrochloride
Lieferant: Thermo Scientific
Synonyme:
Chloropyramine HCl, N1-(4-Chlorobenzyl)-N2,N2-dimethyl-N1-(pyridin-2-yl)ethane-1,2-diamine hydrochloride
J60350.03
J60350.03EA
39.5
EUR
J60350.03
Chloropyramine hydrochloride
Chloropyramine hydrochloride
CAS No.: 6170-42-9
Molecular Formula: C16H20ClN3.HCl
Formula Weight: 326.26
Physical Form: Crystalline powder
Appearance: White
MDL No.: MFCD00079009
Applications: Competitive reversible H-1 receptor antagonist
Formel:
C₁₆H₂₀ClN₃·HCl Molecular Weight: 326,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079009 CAS-Nummer: 6170-42-9 EINECS: 228-216-2 Merck Index: 14,2162 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...