Sodium Cacodylate, 0,1 M Buffer solution, pH 6,5
Lieferant: Thermo Fisher Scientific
J60367.AE
J60367.AEEA
126
EUR
J60367.AE
Sodium Cacodylate, 0,1 M Buffer solution, pH 6,5
Puffer
Biologische Puffer
Liquid.
CAS No.: 6131-99-3
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
MDL No.: MFCD00149079
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...