Ammoniumsulfat 0.5 M in wässriger Lösung

Lieferant: Thermo Fisher Scientific

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

J60368.AD
J60368.ADEA 45 EUR
J60368.AD
Ammoniumsulfat 0.5 M in wässriger Lösung
Ammoniumsulfat

CAS No.: 7783-20-2 Molecular Formula: H8N2O4S Formula Weight: 132.14 Physical Form: Liquid MDL No.: MFCD00003391

Formel: (NH₄)₂SO₄
Molecular Weight: 132.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
EINECS: 231-984-1
Merck Index: 13,00559

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR