Ipratropium bromide 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: rel-(1R,3r,5S,8s)-3-((3-Hydroxy-2-phenylpropanoyl)oxy)-8-isopropyl-8-methyl-8-azabicyclo[3.2.1]octan-8-ium bromide

J60453.MC
J60453.MCEA 106 EUR
J60453.MC
Ipratropium bromide 98%
Ipratropium bromide

Purity: 98% CAS No.: 22254-24-6 Molecular Formula: C20H30BrNO3 Formula Weight: 412.37 Physical Form: Powder Appearance: White Melting Point: 229-231° MDL No.: MFCD00069291


Applications: Muscarinic acetylcholine receptor antagonist and a bronchodilator

Formel: C₂₀H₃₀BrNO₃
Molecular Weight: 412,37 g/mol
Schmelzpunkt: 229…231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00069291
CAS-Nummer: 22254-24-6
EINECS: 244-873-8
Merck Index: 14,5073

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR