Chlorogenic acid
Lieferant: Thermo Fisher Scientific
Synonyme:
(1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-O-Caffeoylquinic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid
CAS No.: 327-97-9
Molecular Formula: C16H18O9
Formula Weight: 354.31
Physical Form: Powder
Appearance: White/pale yellow/beige
Melting Point: 210° dec.
Solubility: Soluble in ethanol, DMSO, or DMF
MDL No.: MFCD00003862
Applications: An analog of caffeic acid
Formel:
C₁₆H₁₈O₉ Molecular Weight: 354.31 g/mol Schmelzpunkt: 210 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003862 CAS-Nummer: 327-97-9 EINECS: 206-325-6 Merck Index: 14,2142 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...