3,3',5,5'-Tetramethylbenzidin (TMB) -Lösung, normale Empfindlichkeit anwendungsbereit, mit gemischter Anzeige
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
TMB
J60461.AP
J60461.APEA
191
EUR
J60461.AP
3,3',5,5'-Tetramethylbenzidin (TMB) -Lösung, normale Empfindlichkeit anwendungsbereit, mit gemischter Anzeige
3,3',5,5'-Tetramethylbenzidin (TMB)
CAS No.: 54827-17-7
Molecular Formula: C16H20N2
Formula Weight: 240.35
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colourless to pale yellow
Beilstein Reference: 2808541
UN No.: UN2810
MDL No.: MFCD00007748
Applications: Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
Formel:
C₁₆H₂₀N₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD00007748 CAS-Nummer: 54827-17-7 EINECS: 259-364-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...