Calpeptin ≥98%
Lieferant: Thermo Scientific
Synonyme:
benzyl N-[(2S)-4-methyl-1-oxo-1-[[(2S)-1-oxohexan-2-yl]amino]pentan-2-yl]carbamate, Cbz-Leu-Nle-al, Benzyl ((S)-4-methyl-1-oxo-1-(((S)-1-oxohexan-2-yl)amino)pentan-2-yl)carbamate
J60481.MA
J60481.MAEA
176
EUR
J60481.MA
Calpeptin ≥98%
Calpeptin
Purity: 98+%
CAS No.: 117591-20-5
Molecular Formula: C20H30N2O4
Formula Weight: 362.50
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in DMSO at 15mg/ml
MDL No.: MFCD00155623
Applications: Inhibitor of the calcium-dependent protease calpain and cathepsin L
Formel:
C₂₀H₃₀N₂O₄ Molecular Weight: 362,47 g/mol |
MDL:
MFCD00155623 CAS-Nummer: 117591-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...