C2 Ceramide 98%
Lieferant: Thermo Scientific
J60534.MA
J60534.MAEA
148
EUR
J60534.MA
C2 Ceramide 98%
C2 Ceramide
Purity: 98%
CAS No.: 3102-57-6
Molecular Formula: C20H39NO3
Formula Weight: 341.53
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 77-78°
Solubility: Soluble in ethanol, methanol, chloroform or DMSO at approximately 10mg/ml
MDL No.: MFCD00153903
Applications: Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases
Formel:
C₂₀H₃₉NO₃ Schmelzpunkt: 77…78 °C |
MDL:
MFCD00153903 CAS-Nummer: 3102-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...