Doxofyllin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
7-(1,3-Dioxolan-2-ylmethyl)theophylline, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethylxanthine, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethyl-1H-purine-2,6-dione
J60575.06
J60575.06EA
126
EUR
J60575.06
Doxofyllin
Doxofylline
CAS No.: 69975-86-6
Molecular Formula: C11H14N4O4
Formula Weight: 266.25
Physical Form: Crystalline powder
Appearance: White
Melting Point: 144-146°
MDL No.: MFCD00865218
Applications: Xanthine bronchodilator that inhibits phosphodiesterase activity
Formel:
C₁₁H₁₄N₄O₄ Molecular Weight: 266,26 g/mol Schmelzpunkt: 144…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00865218 CAS-Nummer: 69975-86-6 EINECS: 274-239-6 Merck Index: 14,3438 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...