Sulphorhodamine 101 acid chloride

Lieferant: Thermo Scientific
Achtung

Synonyme: Texas Red®, Sulphorhodamine 101 sulphonyl chloride

J60581.MA
J60581.MAEA 216 EUR
J60581.MA
Sulphorhodamine 101 acid chloride
Sulphorhodamine 101 acid chloride

CAS No.: 82354-19-6 Molecular Formula: C31H29ClN2O6S2 Formula Weight: 625.15 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Moisture and Light Sensitive Appearance: Dark red Beilstein Reference: 8667894 MDL No.: MFCD00012406


Applications: Fluorescent labeling compound

Formel: C₃₁H₂₉ClN₂O₆S₂
Molecular Weight: 625,151 g/mol
MDL: MFCD00012406
CAS-Nummer: 82354-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR