1,3-Dihydroxyaceton dimer

Lieferant: Thermo Fisher Scientific

J60592.06
J60592.06EA 30.1 EUR
J60592.06
1,3-Dihydroxyaceton dimer
1,3-Dihydroxyaceton dimer

CAS No.: 62147-49-3 Molecular Formula: C3H6O3 Formula Weight: 180.16 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white Melting Point: 75-80° Merck Reference: 14,3182 MDL No.: MFCD00004670

Formel: C₆H₁₂O₆
Schmelzpunkt: 75…80 °C
Dichte: 1,52 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00051019
CAS-Nummer: 62147-49-3
EINECS: 202-494-5
Merck Index: 14,3182

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR