Wasser DNAse_RNAse_free

Lieferant: Thermo Fisher Scientific

Synonyme: Wasserstoffoxid, Hydrogenoxid

J60610.XC
J60610.XCEA 97 EUR
J60610.XC
Wasser DNAse_RNAse_free
Wasser

High quality.


  • Used in all molecular biology applications
  • Suitable for all RNA work
  • No toxic agents in purification procedures


RNase/DNase-free water is used in all molecular biology applications like PCR, DNA. Diethylpyrocarbonate (DEPC)-treated water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work. Toxic agents such as DEPC are not used in purification procedures.

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100 °C
Schmelzpunkt: 0
Dichte: 1.000
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 14,10039

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR