Dihydroergocristine methanesulphonate

Lieferant: Thermo Scientific
Achtung

Synonyme: Dihydroergocristine mesylate

J60638.03
J60638.03EA 405 EUR
J60638.03
Dihydroergocristine methanesulphonate
Dihydroergocristine methanesulphonate

CAS No.: 24730-10-7 Molecular Formula: C36H45N5O8S Formula Weight: 707.80 Physical Form: Powder Appearance: White MDL No.: MFCD00153792


Applications: 5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors

Formel: C₃₆H₄₅N₅O₈S
Molecular Weight: 707,843 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153792
CAS-Nummer: 24730-10-7
EINECS: 246-434-6
Merck Index: 14,3653

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR